Learn More
R&L Enterprises™ Retort Stand Clamps
Enables articles from 2 to 90mm in diameter to be clamped securely. R&L Enterprises™ Retort Stand Clamps feature aluminum alloy jaws with other parts of plated brass.
933.00 NOK - 995.00 NOK
Specifications
Quantity | 5/Pk. |
---|---|
Type | Clamp Retort Stand |
Material | Aluminum Alloy, Brass |
For Use With (Equipment) | For use with 2 to 90mm diameter |
Jaw Type | Cork Lined |
Product Code | Brand | Includes | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | ||||||
11517722
|
R&L Enterprises™
9505 |
Metal headed clamping screw |
995.00 NOK
Pack of 5 |
Estimated Shipment: 17-07-2025
Log in to see stock. |
||||||
11888203
|
R&L Enterprises™
9505/P |
Plastic headed clamping screw |
933.00 NOK
Pack of 5 |
Estimated Shipment: 05-08-2025
Log in to see stock. |
||||||
Description
Enables articles from 2 to 90mm in diameter to be clamped securely. R&L Enterprises™ Retort Stand Clamps feature aluminum alloy jaws with other parts of plated brass.
- Design ensures a firm grip at all angles of opening
- Cork liners for the jaws are well secured
- Available with either metal or plastic clamping screw head
Note:
The clamp and boss head are sold separately.
Specifications
5/Pk. | |
Aluminum Alloy, Brass | |
Cork Lined |
Clamp Retort Stand | |
For use with 2 to 90mm diameter |