Learn More
Restek™ HPLC Stainless Steel Capillary Tubing
Specifications
Type HPLC Capillary Tubing
Finish Smooth
For Use With (Application) HPLC Systems
Diameter (English) Outer 0.06 in.
Diameter (Metric) Outer 0.15 cm
View More Specs
Products 3
Product Code Brand Color Diameter (English) Inner Overall Length (Metric) Price Quantity & Availability  
Product Code Brand Color Diameter (English) Inner Overall Length (Metric) Price Quantity & Availability  
12720580
View Documents
Restek™
25248
Blue 0.01 in. 5 cm N/A
N/A
12730580
View Documents
Restek™
25249
Blue 0.01 in. 10 cm N/A
N/A
12760580
View Documents
Restek™
25253
Yellow 0.02 in. 10 cm N/A
N/A
Specifications
HPLC Capillary Tubing
HPLC Systems
0.15 cm
Smooth
0.06 in.
316 Stainless Steel