Learn More
uvex Respirator Face Mask
Shop All Uvex Products
Click to view available options
Model:
3220 silv-Air Classic
Classic cup style FFP3
silv-Air c 2200 FFP2
silv-Air c 3310 FFP3
Unit Size:
Each
Pack of 15
Pack of 20
Description
- Preformed mask
- Labelled for use with high levels of dust exposure (D)
Specifications
Specifications
Certifications/Compliance (Europe) | EN 149:2001 + A1:2009 |
Color | White |
Design Type | Flat-fold |
Disposable | Single-use |
Filter Type | FFP2 |
Material | Polypropylene, Polyester |
Product Type | Particulate Respirator |
Size | Regular |
With Exhalation Valve | Yes |
Filter Efficiency | FFP2 |
Show More |
Documents
Scientific Resources
Brochures
CE_15395792.pdf
File size: 118 kB
Product Content Correction
Product Title
uvex Respirator Face Mask >