Learn More
Thermo Scientific™ Replacement Stopcock Assembly for Nalgene™ Separatory Funnels
Maintain separatory funnels with this Replacement Stopcock Assembly for Thermo Scientific™ Nalgene™ Separatory Funnels, which ensures a leakproof†fit without lubrication.
Brand: Thermo Scientific™ 714301-1050
1204.96 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Maintain separatory funnels with this Replacement Stopcock Assembly for Thermo Scientific™ Nalgene™ Separatory Funnels, which ensures a leaktight fit without lubrication. This tetrafluoroethylene assembly has excellent chemical resistance and can be chemically disinfected.
- For use with Nalgene™ Separatory Funnels made with Teflon™ fluoropolymer
- Can be chemically sterilized
Specifications
Plastic | |
Replacement Stopcock Assembly; Stopcock Assy TFE 4301-2000 2 L | |
Nalgene Separatory Funnels | |
1/Pk.; 1/Cs. |
Teflon™ is a trademark of The Chemours Company FC, LLC used under license by Nalge Nunc International Corporation.