Learn More
Fisherbrand™ Traceable™ Replacement Printing Paper
For use in Traceable™ Printing Thermohygrometers
150.00 NOK
Specifications
Material | Paper |
---|---|
Ruled | No |
Width (English) Page | 1.5 in. |
Color | White |
For Use With (Equipment) | Traceable™ Printing Thermohygrometers |
Description
- For use in Traceable™ Printing Thermohygrometers
Specifications
Paper | |
1.5 in. | |
Traceable™ Printing Thermohygrometers |
No | |
White | |
Printer Paper |
Product Title
Fisherbrand™ Traceable™ Replacement Printing Paper