Learn More
Masterflex™ C/L™ Power supply for VDC Powered Variable-Speed Pumps; 100 to 240 VAC
Safely adapt your VDC powered C/L™ pump
Brand:  Masterflex™ 77120-11
2647.74 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 13408106
  • 2830.00 NOK / Each
Only null left
Estimated Shipment: 30-07-2025
Description
Suitable for use with C/L™ pumps
Specifications
Specifications
Replacement Power Supply Accessory
115/230 V
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Masterflex™ C/L™ Power supply for VDC Powered Variable-Speed Pumps; 100 to 240 VAC > Voltage: 115/230 VAC