Learn More
Thermo Scientific™ Replacement Needles for GC Syringes
Thermo Scientific™ Replacement Needles for Syringes are available for syringes with removable needles.
Brand:  Thermo Scientific™ 365RN362
644.96 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10243033
  • 685.00 NOK / Pack of 2
Description
Thermo Scientific™ Replacement Needles for Syringes are available for syringes with removable needles.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
365RN362
1.9685 in.
10 ÎĽL
Cone
Removable
26 G
Replacement Syringe Needle
50 mm
Thermo Scientific Model 365D1841 Syringe
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Replacement Needles for GC Syringes > For use with Thermo Scientific syringe 365D1841