Learn More
Thermo Scientific™ Replacement Microcentrifuge Rotor Lids
Combine these replacement microcentrifuge rotor lids with the microcentrifuge rotor.
Brand:  Thermo Scientific™ 75003410
3838.11 NOK valid until 2025-07-13
Use promo code "24135" to get your promotional price.
 View more versions of this product
Product Code. 10177222
  • 4430.57 NOK / Each
Spotlight
Use promo code 24156 at checkout
View Available Promotions
Edge
Description
Description
Combine Thermo Scientific™ Replacement Microcentrifuge Rotor Lids with the microcentrifuge rotor.
ClickSeal™ Biocontainment Lid:
  • For use with 24 Ă— 1.5/2.0 mL Rotor (75003424), PCR 4 Ă— 8 Rotor (75003440)
  • Unique design clicks when proper seal is made, providing assurance that your samples are securely contained, as well as shortens retrieval time with one click opening
  • With PPSU rotor lid, view contents of rotor for added safety and convenience
Screw-on Lid:
  • For use with 36 Ă— 0.5 mL Rotor (75003436), Dual Row 18 Ă— 2.00.5 mL Rotor (75003418), PCR 8 Ă— 8 Rotor (75003489)
Ordering Information:
Sold individually.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ClickSeal Lid
24 x 1.5/2 mL Rotor (75003424), PCR 4 x 8 Rotor
ClickSeal™ Biocontainment Lid
Certificates
View accessory products
Promotions
Promotions
  • Multiple Manufacturers
    Save up to 59% on some of our most popular products
    Promo Code: 24140
    Expires2025-08-24
  • Multiple Manufacturers
    Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.
    Promo Code: 24156
    Expires2025-06-30
    Hot Deal
    *Discount will apply to qualifying orders received by Fisher Scientific between 01/05/2025 and 30/06/2025. Promotion code must be presented at the time of order. Discount applies to the list price in effect at the time the order is received. Promotion cannot be combined with other discounts or promotions. Offer void where prohibited, licensed, or restricted by federal, state, provincial, or local laws or regulation or agency/institutional policy. Other restrictions may apply.
    Terms & Conditions See Full Promo Details
  • Multiple Manufacturers
    Get amazing savings on products for your analyses
    Promo Code: 24135
    Expires2025-07-13
  • Multiple Manufacturers
    Browse exclusive prices for our academic customers
    Promo Code: 25081
    Expires2025-12-31
  • Multiple Manufacturers
    Save big on essential products for cancer research
    Promo Code24147
    Expires2025-07-31
Product Content Correction
Product Title
Thermo Scientific™ Replacement Microcentrifuge Rotor Lids > Replacement ClickSeal Biocontainment Lid