Learn More
Saint-Gobain Replacement Blade
Use this blade with benchtop tubing cutters to cut tubing with an outer diameter of 1/16 to 5/8 in.
Brand:  Saint-Gobain 06438-11
Product Code. 11771429
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Replacement blade for benchtop tubing cutter
  • Steel blade makes right-angle cuts in plastic tubing
Specifications
Specifications
Replacement Blade
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Saint-Gobain Replacement Blade > Replacement blade