Learn More
Merck Replacement Battery
MilliporeSigma Replacement Battery, Voltage: 6V, Battery Type: Replacement Battery, Quantity: 1Each
Brand: Merck MERSBAT01
This item is not returnable.
View return policy
Description
MilliporeSigma Replacement Battery, Voltage: 6V, Battery Type: Replacement Battery, Quantity: 1Each
Specifications
Replacement Battery | |
1 Ea. | |
6 V |
Millicell Inserts and Plates
File size: 1.4 MB
Product Content Correction
Product Title
Merck Replacement Battery > Voltage: 6V