Learn More
Grant Instruments™ Replacement 1L bottle with blue cap, insert and sealer
Center hole accommodates aspirator tubing. Grant Instruments Replacement Blue Cap for 1L Bottle includes seal and insert.
Brand: Grant Instruments™ FTA-B
354.35 NOK valid until 2025-07-11
Use promo code "25721" to get your promotional price.
Description
- Center hole accommodates aspirator tubing
- Fits 1L bottle
- Cap includes insert and seal
Specifications
FTA-1 Aspirator |
Special Offers
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now