Learn More
Trajan Scientific and Medical™ Removable Needle for Agilent Autosampler Syringes
Quality needle replacements for use with Agilent autosampler syringes with removable needle.
485.00 NOK
Specifications
Product Type | Replacement Needle |
---|---|
Needle Point Style | Cone |
For Use With (Equipment) | Syringes (10ÎĽL) |
Product Code | Brand | Needle Gauge | Diameter (Metric) Outer | Length (Metric) Needle | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Needle Gauge | Diameter (Metric) Outer | Length (Metric) Needle | Price | Quantity & Availability | |||
10258770
|
Trajan Scientific and Medical™
037715 |
26 G | 0.47 mm | 42 mm |
485.00 NOK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10075620
|
Trajan Scientific and Medical™
037717 |
23 G | 0.63 mm | 50 mm |
485.00 NOK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Needle length is the exposed length of needle
- For use with Agilent Technologies autosamplers
Specifications
Replacement Needle | |
Cone | |
Syringes (10ÎĽL) |
Product Title
Trajan Scientific and Medical™ Removable Needle for Agilent Autosampler Syringes