Learn More
Thermo Scientific™ Remel™ McFarland Equivalence Turbidity Standards
Thermo Scientific™ Remel McFarland Equivalence Turbidity Standard is packaged in plastic case with a visual comparison card.
Brand:  Thermo Scientific™ R20411
481.71 NOK valid until 2025-04-30
Use promo code "24235" to get your promotional price.
 View more versions of this product
Product Code. 10037212
  • 550.00 NOK / Each
Explore more special offers
Description
Description
Thermo Scientific™ Remel McFarland Equivalence Turbidity Standard is packaged in plastic case with a visual comparison card.
  • Tube size is 15 x 103mm and fits in most spectrophotometers
  • Polystyrene technology allows longer shelf life
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Turbidity Standard 1.0
15 x 103 mm
Documents
Instructions
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6212743 Certificate TypeCertificate of Analysis Date25/03/2025 Catalog NumberR20411
Lot Number6212742 Certificate TypeCertificate of Analysis Date28/02/2025 Catalog NumberR20411
Lot Number6185620 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberR20411
Lot Number6175942 Certificate TypeCertificate of Analysis Date19/11/2024 Catalog NumberR20411
Lot Number6164226 Certificate TypeCertificate of Analysis Date18/10/2024 Catalog NumberR20411
Special Offers
Special Offers
  • Multiple Manufacturers
    Discover exclusive monthly offers on scientific equipment to enhance your laboratory's capabilities.
    Promo Code: 24235
    Expires2025-04-30
Product Content Correction
Product Title
Thermo Scientific™ Remel™ McFarland Equivalence Turbidity Standards > Turbidity Standard 1.0