Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, Haemophilus influenzae
Used for serological identification of H. influenzae by rapid slide agglutination test and CIE procedures.
Brand:  Thermo Scientific™ R30166301
3602.89 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 12643528
  • 3700.00 NOK / Each
Explore more special offers
Description
Description
Remel Agglutinating Sera are used for serological identification of H. influenzae by rapid slide agglutination test and CIE procedures.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Agglutinating Sera, Haemophilus influenzae Type D
H. influenzae
Agglutinating Sera
Documents
Instructions
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6127644 Certificate TypeCertificate of Analysis Date06/11/2024 Catalog NumberR30166301
Lot Number3800013 Certificate TypeCertificate of Analysis Date06/04/2024 Catalog NumberR30166301
Lot Number3753271 Certificate TypeCertificate of Analysis Date07/02/2024 Catalog NumberR30166301
Lot Number3671601 Certificate TypeCertificate of Analysis Date21/10/2023 Catalog NumberR30166301
Lot Number3644900 Certificate TypeCertificate of Analysis Date26/05/2023 Catalog NumberR30166301
Product Title
Thermo Scientific™ Remel™ Agglutinating Sera, Haemophilus influenzae > Type D