Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, E. coli Polyvalent
Used for serological identification of E. coli by slide and/or tube agglutination tests.
Brand:  Thermo Scientific™ R30955001
1246.76 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 12637539
  • 1280.00 NOK / Each
Description
Remel Agglutinating Sera, E. coli Polyvalent is used for serological identification of E. coli by slide and/or tube agglutination tests.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
E. coli Polyvalent 3 (types O86, O114, O125, O127, O128)
Agglutinating Sera
2 mL
E. coli
2 mL/Vial
Documents
Instructions
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3702136 Certificate TypeCertificate of Analysis Date30/07/2024 Catalog NumberR30955001
Lot Number3605834 Certificate TypeCertificate of Analysis Date24/03/2023 Catalog NumberR30955001
Lot Number3544012 Certificate TypeCertificate of Analysis Date20/09/2022 Catalog NumberR30955001
Lot Number3412899 Certificate TypeCertificate of Analysis Date21/02/2022 Catalog NumberR30955001
Lot Number3298147 Certificate TypeCertificate of Analysis Date10/11/2021 Catalog NumberR30955001
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Remel™ Agglutinating Sera, E. coli Polyvalent > E. coli Polyvalent 3 (types O86, O114, O125, O127, O128)