Learn More
Kautex™ Red Polypropylene Screw Closures
Seal a container's ingredients with these Kautex™ Red Polypropylene Screw Closures when you package, send samples, transport and store their contents.
Brand:  Kautex™ 2000083762
 View more versions of this product
Product Code. 10772761
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • PP
  • Red
  • Tamper evident ring
  • PE-foam insert
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Red
Polypropylene
Screw Cap
80 mm
Tamper Evident Ring and Polyethylene Foam Insert
Polyethylene Foam Insert
1500/2500/4000 mL Bottle
SDS
Documents Certificates
Promotions
Promotions
Product Content Correction
Product Title
Kautex™ Red Polypropylene Screw Closures > For Use With: 1500/2500/4000 mL Bottle