Learn More
Helios Italquartz Rectangular Basins/Dishes, Transparent
Rectangular basins/dishes constructed of transparent, durable, resistant, and chemically inert quartz glass.
Brand: Helios Italquartz 62P00206
This item is not returnable.
View return policy
Description
- Transparent
- Rectangular form
- Chemically inert
- Excellent electrical insulation
- Excellent strength and durability
- Exceptional thermal stability compared to glass
Specifications
Transparent | |
Rectangular | |
115 x 12 mm | |
12 mm |
Quartz | |
Dish | |
115/60 mm | |
12 mm |
Product Content Correction
Product Title
Helios Italquartz Rectangular Basins/Dishes, Transparent > 115 x 12 mm