Learn More
Helios Italquartz Rectangular Basins/Dishes, Transparent
Rectangular basins/dishes constructed of transparent, durable, resistant, and chemically inert quartz glass.
Brand:  Helios Italquartz 62P00205
 View more versions of this product
Product Code. 15808044
  • 2030.00 NOK / Each
Only null left
Estimated Shipment: 05-08-2025
Add to basket
Description
Description
  • Transparent
  • Rectangular form
  • Chemically inert
  • Excellent electrical insulation
  • Excellent strength and durability
  • Exceptional thermal stability compared to glass
Specifications
Specifications
Transparent
Rectangular
68 x 12 mm
12 mm
Quartz
Dish
68/39 mm
12 mm
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Helios Italquartz Rectangular Basins/Dishes, Transparent > 68 x 12 mm