Learn More
R&D Systems™ Recombinant Rat Erythropoietin/EPO Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 1306-RE-010
3052.19 NOK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The Recombinant Rat Erythropoietin Protein is derived from Sf 21 (stably transfected). The Recombinant Rat Erythropoietin Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. | |
18.5 kDa | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
ECYT5, EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2 | |
Recombinant |
24335 | |
Erythropoietin/EPO | |
Spodoptera frugiperda, Sf 21 (stably transfected)-derived rat Erythropoietin/EPO protein Ala27-Arg192 | |
<0.10 EU per 1μg of the protein by the LAL method. | |
Unconjugated | |
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |