missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse LOXL2 / Lysyl oxidase homolog 2 Protein
A DNA sequence encoding the Mus musculus (Mouse) LOXL2 / Lysyl oxidase homolog 2, was expressed in the hosts and tags indicated.
2526.25 NOK - 38140.00 NOK
Specifications
Accession Number | NP_201582.2 |
---|---|
Gene ID (Entrez) | 94352 |
Name | LOXL2 / Lysyl oxidase homolog 2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16042430
|
enQuireBio™
QP6317-ec-10ug |
10 μg | N/A | ||||||
16082430
|
enQuireBio™
QP6317-ec-50ug |
50 μg | N/A | ||||||
16032430
|
enQuireBio™
QP6317-ec-100ug |
100 μg |
8590.00 NOK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16062430
|
enQuireBio™
QP6317-ec-200ug |
200 μg | N/A | ||||||
16072430
|
enQuireBio™
QP6317-ec-500ug |
500 μg | N/A | ||||||
16052430
|
enQuireBio™
QP6317-ec-1mg |
1 mg |
38140.00 NOK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
NP_201582.2 | |
LOXL2 / Lysyl oxidase homolog 2 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
94352 | |
Research Use Only | |
Loxl2 | |
Recombinant Protein | |
E. coli | |
QYEGWPYQLQ YPEYFQQPAP EHHQRQVPSD VVKIQVRLAG QKRKHNEGRV EVYYEGQWGT VCDDDFSIHA AHVVCRQVGY VEAKSWAASS SYGPGEGPIW LDNIYCTGKE STLASCSSNG WGVTDCKHTE DVGVVCSEKR IPGFKFDNSL INQIESLNIQ VEDIRIRPIL SAFRHRKPVT EGYVEVKEGK AWKQICNKHW TAKNSHVVCG MFGFPAEKTY NPKAYKTFAS RRKLRYWKFS MNCTGTEAHI SSCKLGPSVT RDPVKNATCE NGQPAVVSCV PSQIFSPDGP SRFRKAYKPE QPLVRLRGGA QVGEGRVEVL KNGEWGTICD DKWDLVSASV VCRELGFGTA KEAITGSRLG QGIGPIHLNE VQCTGTEKSI IDCKFNTESQ GCNHEEDAGV RCNIPIMGFQ KKVRLNGGRN PYEGRVEVLT ERNGSLVWGT VCGQNWGIVE AMVVCRQLGL GFASNAFQET WYWHGNIFAN NVVMSGVKCS GTELSLAHCR HDEEVACPEG GVRFGAGVAC SETAPDLVLN AEIVQQTAYL EDRPMSLLQC AMEENCLSAS AVHTDPTRGH RRLLRFSSQI HNNGQSDFRP KNGRHAWIWH DCHRHYHSME VFTYYDLLSL NGTKVAEGHK ASFCLEDTEC EGDIQKSYEC ANFGEQGITM GCWDMYRHDI DCQWIDITDV PPGDYLFQVV INPNYEVPES DFSNNIMKCR SRYDGYRIWM YNCHVGGAFS EETEQKFEHF SGLLNNQLSV Q | |
Greater than 90% as determined by SDS-PAGE. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Mouse LOXL2 / Lysyl oxidase homolog 2 Protein