missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human USH1C His Protein
Highly purified. Generating reliable and reproducible results. Applications: SDS-Page
9495.00 NOK
Specifications
Accession Number | AAH16057 |
---|---|
For Use With (Application) | ELISA |
Formulation | 20mM Tris-HCl buffer (pH 8.0), 20% glycerol |
Gene ID (Entrez) | 10083 |
Molecular Weight (g/mol) | 64.6kDa |
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-533 of Human USH1C The Recombinant Human USH1C Protein is derived from E. coli. The Recombinant Human USH1C Protein has been validated for the following applications: SDS-Page.Specifications
AAH16057 | |
20mM Tris-HCl buffer (pH 8.0), 20% glycerol | |
64.6kDa | |
Protein | |
-80°C. Avoid freeze-thaw cycles. | |
>95%, by SDS-PAGE |
ELISA | |
10083 | |
USH1C Protein | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSHMDR KVAREFRHKV DFLIENDAEK DYLYDVLRMY HQTMDVAVLV GDLKLVINEP SRLPLFDAIR PLIPLKHQVE YDQLTPRRSR KLKEVRLDRL HPEGLGLSVR GGLEFGCGLF ISHLIKGGQA DSVGLQVGDE IVRINGYSIS SCTHEEVINL IRTKKTVSIK VRHIGLIPVK SSPDEPLTWQ YVDQFVSESG GVRGSLGSPG NRENKEKKVF ISLVGSRGLG CSISSGPIQK PGIFISHVKP GSLSAEVGLE IGDQIVEVNG VDFSNLDHKE GRELFMTDRE RLAEARQREL QRQELLMQKR LAMESNKILQ EQQEMERQRR KEIAQKAAEE NERYRKEMEQ IVEEEEKFKK QWEEDWGSKE QLLLPKTITA EVHPVPLRKP KYDQGVEPEL EPADDLDGGT EEQGEQDFRK YEEGFDPYSM FTPEQIMGKD VRLLRIKKEG SLDLALEGGV DSPIGKVVVS AVYERGAAER HGGIVKGDEI MAINGKIVTD YTLAEADAAL QKAWNQGGDW IDLVVAVCPP KEYDDELTFF | |
Human |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Recombinant Human USH1C His Protein