Learn More
R&D Systems™ Recombinant Human TLR4 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 1478-TR-050
4042.22 NOK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
The Recombinant Human TLR4 Protein is derived from NS0. The Recombinant Human TLR4 Protein has been validated for the following applications: Bioactivity.Specifications
Lyophilized from a 0.2μm filtered solution in PBS. | |
70.6 kDa | |
50 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
7099 | |
TLR4 | |
Mouse myeloma cell line,NS0-derived human TLR4 protein Glu24-Lys631,with a C-terminal Ser and 10-His tag | |
<0.1 EU per 1μg of the protein by the LAL method. | |
Recombinant |