missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TGF-beta 1 / TGFB1 Protein

A DNA sequence encoding the Homo sapiens (Human) TGF-beta 1 / TGFB1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8568-ec-10ug

 View more versions of this product

Product Code. 16075331

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

NP_000651.3
TGF-beta 1 / TGFB1 Protein
Research Use Only
TGFB1
Recombinant Protein
E. coli
DTNYCFSSTE KNCCVRQLYI DFRKDLGWKW IHEPKGYHAN FCLGPCPYIW SLDTQYSKVL ALYNQHNPGA SAAPCCVPQA LEPLPIVYYV GRKPKVEQLS NMIVRSCKCS
Greater than 90% as determined by SDS-PAGE.
7040
10 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers