missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Tektin-5 Protein
A DNA sequence encoding the Homo sapiens (Human) Tektin-5, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8007-ec-50ug
This item is not returnable.
View return policy
Specifications
NP_653275.1 | |
Tektin-5 Protein | |
Research Use Only | |
TEKT5 | |
Recombinant Protein | |
E. coli | |
MEFLGTTQTA SYCGPKKCCG LTSLPAVQAP VIQECYQPYY LPGYRYLNSW RPSLFYKIAN VQTCPDESTS TLRPPTILPT LRSALFSRYS PHDWDQSNQL QVRGAEASRL WASRLTDDSM RLLQDKDQLT HQMQEGTCRN LGQRLSDIGF WKSELSYELD RLLTENQNLE TVKRRLECAA NEVNCPLQVA LECLYHREKR IGIDLVHDNV EKNLIREVDL LKCCQEQMRK LAQRIDIQMR DNRDAQHVLE RDLEDKSSAQ CIDEKCFNLR NTSDCISFFH GMEKIDGTIS VPETWAKFSN DNIKHSQNMR ANSIQLREEA EHLFETLSDQ MWRQFTDTNL AFNARISEVT DVKNKLQTQL AKTLQEIFQA ENTIMLLERS IMAKEGPLKV AQTRLECRTR RPNMELCRDI PQLKLVNEVF TIDDTLQTLK LRLRETQDTL QLLVMTKCRL EHELAIKANT LCIDKEKCMG MRKTFPCTPR LVGHT | |
Greater than 90% as determined by SDS-PAGE. |
146279 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
enQuireBio™ Recombinant Human Tektin-5 Protein > 50μg
Spot an opportunity for improvement?Share a Content Correction