missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human RTP1 GST (N-Term) Protein
Recombinant Protein
Brand: Novus Biologicals™ H00132112-Q01-10ug
This item is not returnable.
View return policy
Description
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
Specifications
Western Blot, ELISA, Protein Array, Immunoaffinity Purification | |
132112 | |
10 μg | |
MGC35450, receptor (chemosensory) transporter protein 1, receptor transporter protein 1, receptor transporting protein 1, receptor-transporting protein 1 | |
>80% by SDS-PAGE and Coomassie blue staining |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. | |
MolecularWeight-theroretical: 35.64 kDa | |
Store at -80°C. Avoid freeze/thaw cycles. | |
RTP1 | |
RTP1 |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 7
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Recombinant Human RTP1 GST (N-Term) Protein > 10 μg
Spot an opportunity for improvement?Share a Content Correction