missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human MKP-1/DUSP1 GST (N-Term) Protein
Recombinant Protein
5645.00 NOK - 7395.00 NOK
Specifications
For Use With (Application) | Western Blot, ELISA, Protein Array, Immunoaffinity Purification |
---|---|
Formulation | 50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. |
Gene ID (Entrez) | 1843 |
Molecular Weight (g/mol) | M.W. (Theoretical): 32.67 kDa |
Storage Requirements | Store at -80C. Avoid freeze-thaw cycles. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18376567
|
Novus Biologicals™
H00001843-Q01-10ug |
10 μg |
5645.00 NOK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18386217
|
Novus Biologicals™
H00001843-Q01-25ug |
25 μg |
7395.00 NOK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.Specifications
Western Blot, ELISA, Protein Array, Immunoaffinity Purification | |
1843 | |
Store at -80C. Avoid freeze-thaw cycles. | |
DUSP1 | |
>80% by SDS-PAGE and Coomassie blue staining |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. | |
M.W. (Theoretical): 32.67 kDa | |
CL100MAP kinase phosphatase 1, dual specificity phosphatase 1, Dual specificity protein phosphatase hVH1, EC 3.1.3.16, EC 3.1.3.48, HVH1, Mitogen-activated protein kinase phosphatase 1, MKP1, MKP-1dual specificity protein phosphatase 1, Protein-tyrosine phosphatase CL100, PTPN10serine/threonine specific protein phosphatase, VH1 | |
Cell Cycle and Replication | |
MKP-1/DUSP1 |