Learn More
R&D Systems™ Recombinant Human LBP Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Brand: R&D Systems™ 870-LP-025
4233.15 NOK valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The Recombinant Human LBP Protein is derived from NS0. The Recombinant Human LBP Protein has been validated for the following applications: Bioactivity.Spezifikation
Lyophilized from a 0.2μm filtered solution in Tris, NaCl and EDTA with BSA as a carrier protein. | |
Predicted MW: 51kDa Observed MW: 55-60kDa, reducing conditions | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
3929 | |
LBP | |
Mouse myeloma cell line,NS0-derived human LBP protein Ala26-Val481 | |
Unconjugated | |
97%, by SDS-PAGE under reducing conditions and visualized by silver stain. |