missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Human DGCR14 GST (N-Term) Protein
Recombinant Protein
Brand: Novus Biologicals™ H00008220-P01-25ug
This item is not returnable.
View return policy
Description
This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.Specifications
Western Blot, ELISA, PAGE, Protein Array, Immunoaffinity Purification | |
8220 | |
25 μg | |
DGCR13, DGS-HDGS-I, DGSIDGSH, DiGeorge syndrome critical region 13, DiGeorge syndrome critical region 14, DiGeorge syndrome critical region gene 13, DiGeorge syndrome critical region gene 14, DiGeorge syndrome gene H, DiGeorge syndrome gene I, DiGeorge syndrome protein H, ES2DiGeorge syndrome critical region gene DGSI, Es2el, Protein DGCR13, protein DGCR14, Protein ES2 | |
Neuroscience | |
DGCR14 |
50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer. | |
67 kDa | |
Store at -80°C. Avoid freeze-thaw cycles. | |
ESS2 | |
>80% by SDS-PAGE and Coomassie blue staining |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction