Learn More
R&D Systems™ Recombinant Human CD39L3/ENTPD3 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Brand: R&D Systems™ 4400-EN-010
This item is not returnable.
View return policy
Description
The Recombinant Human CD39L3/ENTPD3 Protein is derived from NS0. The Recombinant Human CD39L3/ENTPD3 Protein has been validated for the following applications: Enzyme Activity.Specifications
Supplied as a 0.2 μm filtered solution in Tris, CaCl 2 NaCl and Glycerol. | |
50 kDa | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening. | |
Recombinant |
956 | |
CD39L3/ENTPD3 | |
Mouse myeloma cell line,NS0-derived human CD39L3/ENTPD3 protein Gln44-Pro485,with a C-terminal 6-His tag | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |