Learn More
R&D Systems™ Recombinant Human CD39L3/ENTPD3 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Brand:  R&D Systems™ 4400-EN-010
Product Code. 18196440
  • 5535.00 NOK / 10µg
Explore more special offers
Description
Description
The Recombinant Human CD39L3/ENTPD3 Protein is derived from NS0. The Recombinant Human CD39L3/ENTPD3 Protein has been validated for the following applications: Enzyme Activity.
Specifications
Specifications
Supplied as a 0.2 μm filtered solution in Tris, CaCl 2 NaCl and Glycerol.
50 kDa
10 μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 3 months, -20 to -70° C under sterile conditions after opening.
Recombinant
956
CD39L3/ENTPD3
Mouse myeloma cell line,NS0-derived human CD39L3/ENTPD3 protein Gln44-Pro485,with a C-terminal 6-His tag
Unconjugated
95%, by SDS-PAGE under reducing conditions and visualized by silver stain.