missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals Recombinant Human beta-Synuclein Active, Monomer Protein
4830.00 NOK - 15815.00 NOK
Specifications
For Use With (Application) | In vitro Assay, In vivo Assay, SDS-PAGE, Western Blot |
---|---|
Formulation | PBS pH 7.4 |
Gene ID (Entrez) | 6620 |
Storage Requirements | Store at -80C. Avoid freeze-thaw cycles. |
Gene Alias | beta-synuclein, synuclein, beta |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18088335
|
Novus Biologicals
NBP3-14775-100UG |
100 μg |
4830.00 NOK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18315095
|
Novus Biologicals
NBP3-14775-200UG |
2 x 100 μg Vials |
7515.00 NOK
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18329685
|
Novus Biologicals
NBP3-14775-500UG |
5 x 100 μg Vials |
15815.00 NOK
1 set |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
In vitro Assay, In vivo Assay, SDS-PAGE, Western Blot | |
6620 | |
beta-synuclein, synuclein, beta | |
Alzheimers Research, Neuroscience | |
beta-Synuclein |
PBS pH 7.4 | |
Store at -80C. Avoid freeze-thaw cycles. | |
SNCB | |
>95%, by SDS-PAGE |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title