missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Angiopoietin-4 Protein
Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity
5205.00 NOK
Specifications
Gene ID (Entrez) | 51378 |
---|---|
Molecular Weight (g/mol) | 57 kDa (monomer) |
Name | Angiopoietin-4 |
Quantity | 25 μg |
Source | Mouse myeloma cell line,NS0-derived human Angiopoietin-4 protein, (N-terminus) Human Ang-4 (Gln23-Ile503) or (Gln24-Ile503) Accession &Num; Q9Y264 IEGR GGGSGGGSGGGS 10-His tag (C-terminus) |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18394817
|
R&D Systems™
964-AN-025 |
Lyophilized from a 0.2μm filtered solution in Tris-Citrate and NaCl with BSA as a carrier protein. |
5205.00 NOK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18335537
|
R&D Systems™
964-AN-025/CF |
Lyophilized from a 0.2μm filtered solution in Tris-Citrate and NaCl. |
5205.00 NOK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Recombinant Human Angiopoietin-4 Protein is derived from NS0. The Recombinant Human Angiopoietin-4 Protein has been validated for the following applications: Binding Activity.Specifications
51378 | |
Angiopoietin-4 | |
Mouse myeloma cell line,NS0-derived human Angiopoietin-4 protein, (N-terminus) Human Ang-4 (Gln23-Ile503) or (Gln24-Ile503) Accession &Num; Q9Y264 IEGR GGGSGGGSGGGS 10-His tag (C-terminus) | |
Oligomer | |
Recombinant |
57 kDa (monomer) | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
85%, by SDS-PAGE under reducing conditions and visualized by silver stain. |