Learn More
Testo™ Rechargeable Batteries
Brand:  Testo™ 0515 0025
Product Code. 10608771
  • 358.00 NOK / Each
Only null left
Estimated Shipment: 18-09-2025
Specifications
Rechargeable Battery
9 V
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Testo™ Rechargeable Batteries > Voltage: 9V