Learn More
Lenz Laborglasintrumente Receiving Flasks with PUR Coating
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
1100.00 NOK - 1760.00 NOK
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
11903401
|
Lenz Laborglasintrumente
03064073 |
2 L |
1760.00 NOK
Each |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||||
11973391
|
Lenz Laborglasintrumente
03064049 |
250 mL |
1100.00 NOK
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
|||||
Description
- Especially for rotary evaporators
- With spherical joint socket S 35
- With highly transparent PUR coating
- Autoclavable
- Material: DURAN™
Specifications
Duran Glass |