Learn More
Thermo Scientific™ Reagent Reservoirs
Click to view available options
Capacity (Metric):
25 mL
60 mL
100 mL
Material:
Polypropylene
Polystyrene
Sterility:
Non-sterile
Packaging:
10 Bags of 10
Bulk
Unit Size:
Pack of 100
Pack of 5
Description
Thermo Scientific™ Reagent Reservoirs provide a variety of solutions to suit all of your pipetting procedures.
Innovation goes beyond our pipetters and tips. Even our reagent reservoirs have been designed to provide solutions tailored to your procedures, rather than the other way around. Four variations of reservoirs are offered, 25mL, 25mL with divider, 100mL and a disposable polystyrene 75mL reservoir.
Features:
- Trough within a trough maximizes the amount of liquid accessible to pipette tips when using small amounts of reagent
- Pour-off spouts on all four corners reduce spillage when pouring reagents out of reservoir
- Graduations on inside wall enable quick measurement of remaining liquid
- Extra-wide base adds rigidity and stability to reservoir, helping to avoid spills
- 25mL divided reservoir allows the pipetting of two different reagents with up to eight channels on one side, and up to four on the other; maximizes recovery if using only single channel and small amount of reagent
Specifications
Specifications
Color | White |
Capacity (Metric) | 25 mL |
Material | Polystyrene |
Sterility | Non-sterile |
Type | Reagent Reservoir with Divider |
Packaging | 10 Bags of 10 |
Disposable | Single-use |
Product Content Correction
Product Title
Thermo Scientific™ Reagent Reservoirs >