Learn More
Lonza™ Reagent Reservoir
Used with multi-channel pipettors when adding reagents to a 96-well plate. Lonza™ Reagent Reservoir is tested for endotoxin and is certified to meet a specification of <0.005 EU/mL. 10 disposable trays per pack.
Brand: Lonza™ 190035
Description
These disposable reservoirs are provided in a zip closure bag for convenient storage of unused reservoirs until they are ready to be used.Specifications
Endotoxins: <0.005EU/mL | |
Plastic | |
10 Reservoirs |
White | |
Reagent Reservoir | |
Zip-Closure Bag |