Learn More
Thermo Scientific™ Gas Regulators for Reacti-Vap™ Evaporators
Use these gas regulators to deliver the right amount of gas for sample evaporation using the Thermo Scientific™ ReactiVap™ Evaporator.
4060.61 NOK - 5055.00 NOK
Specifications
For Use With Reacti-Vap Evaporators
View More Specs
Products 2
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
15224613
View Documents
Thermo Scientific™
60181-625
Gas Regulator, 1/8in. Tube Fitting
4060.61 NOK
Each
Estimated Shipment: 10-06-2025
Add to basket
15234613
View Documents
Thermo Scientific™
60181-626
Gas Regulator, 1/4in. Tube Fitting
5055.00 NOK
Each
In Stock
Add to basket
Description
Description
Deliver the right amount of gas to your Thermo Scientific™ ReactiVap™ Evaporator using these gas regulators. The Reacti-Vap Evaporator delivers pressurized gas to easily and efficiently evaporate samples for gas chromatography (GC) analysis. Choose between gas regulators with a 1/8 inch or a 1/4 inch tube fitting.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Specifications
Reacti-Vap Evaporators
Documents
Product Certifications
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Product Content Correction
Product Title
Thermo Scientific™ Gas Regulators for Reacti-Vap™ Evaporators
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Merci de nous aider à améliorer notre site web. Votre commentaire a été soumis