Learn More
Thermo Scientific™ Reacti-Therm™ Thermometers
Use these thermometers to measure temperature in dry block heaters of Thermo Scientific™ Reacti-Therm™ Heating and Stirring module.
Brand:  Thermo Scientific™ TS-18915
1577.23 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 11547510
  • 1665.00 NOK / Each
Only null left
In Stock
Description
Measure temperature in dry block heaters of the Thermo Scientific™ Reacti-Therm™ Heating and Stirring module and other laboratory equipment using Thermo Scientific™ Reacti-Therm™ Thermometers. The thermometers are mercury-free and alcohol-filled for greater safety. PTFE coating ensures that glass is impervious to corrosive materials. They are available in two different temperature ranges.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Thermometer
0°C to 200°C
Dry Block Heaters and Other Instruments
SDS
Documents Certificates
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Reacti-Therm™ Thermometers > 0° to 200°C