missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RCL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15330-1-AP-150UL
This item is not returnable.
View return policy
Description
RCL1, which does not have cyclase activity, plays a role in 40S- ribosomal-subunit biogenesis in the early pre-rRNA processing steps at sites A0, A1 and A2 that are required for proper maturation of the 18S RNA.Specifications
RCL1 | |
Polyclonal | |
Unconjugated | |
RCL1 | |
RCL1, RNAC, RPC2, RPCL1, RTC2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10171, 309301, 59028 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9JJT0, Q9Y2P8 | |
Rcl1 | |
RCL1 Fusion Protein Ag7058 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction