missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBX1 Mouse anti-Human, Mouse, Rat, Clone: 1F4D9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66716-1-IG-20UL
This item is not returnable.
View return policy
Description
This locus encodes a RING finger-like domain-containing protein. The encoded protein interacts with cullin proteins and likely plays a role in ubiquitination processes necessary for cell cycle progression. This protein may also affect protein turnover. Related pseudogenes exist on chromosomes 2 and 5.Specifications
RBX1 | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P62877, P62878 | |
Rbx1 | |
RBX1 Fusion Protein Ag7005 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG1 |
Western Blot, Immunohistochemistry (Paraffin) | |
1F4D9 | |
Unconjugated | |
RBX1 | |
BA554C12.1, Protein ZYP, RBX1, Regulator of cullins 1, ring box 1, RING box protein 1, RING finger protein 75, RNF75, ROC1 | |
Mouse | |
Protein G | |
RUO | |
300084, 56438, 9978 | |
-20°C | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction