missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM41 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1696.32 NOK - 4796.50 NOK
Specifications
Antigen | RBM41 |
---|---|
Concentration | 0.17 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16895114
|
Proteintech
15784-1-AP-20UL |
20 μL |
1696.32 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16885114
|
Proteintech
15784-1-AP-150UL |
150 μL |
4796.50 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
May bind RNA.Specifications
RBM41 | |
Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q8JZV4, Q96IZ5 | |
237073, 55285, 680581 | |
RBM41 Fusion Protein Ag8483 | |
Primary | |
-20°C | |
RBM41 |
0.17 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
bB383K5.1, RBM41, RNA binding motif protein 41, RNA binding protein 41 | |
RBM41 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title