missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ravulizumab Recombinant Human Monoclonal Antibody, Invitrogen™
Human Recombinant Monoclonal Antibody
Brand: Invitrogen MA542134
This item is not returnable.
View return policy
Description
Endotoxin level < 0.01EU/μg by LAL method Use a manual defrost freezer and avoid repeated freeze thaw cycles. Store at 2 to 8 °C for one week. Store at -20 to -80 °C for twelve months from the date of receipt.
Ravulizumab is considered a long-acting complement 5 (C5) inhibitor that has been undergoing clinical trials for the treatment of paroxysmal nocturnal hemoglobinuria (PNH) as of 4 February, 2016. A pharmaceutical similar to ravulizumab (ALXN1210), called eculizumab, is currently approved for the treatment of PNH in 46 countries under the brand name Soliris®. Ravulizumab is considered by Alexion Pharmaceuticals Inc. to be a "e;next-generation"e; eculizumab molecule. Ravulizumab was subsequently approved by the US FDA in December of 2018 for a variety of beneficial characteristics that make it an advanced, next-generation agent in comparison to eculizumab. In particular, ravulizumab is currently the first and only long-acting C5 complement inhibitor that can be administered every eight weeks for the treatment of adult patients with PNH whereas eculizumab is a bi-weekly treatment, Label. Moreover, virtually all of the phase 3 trial results for ravulizumab have demonstrated the equivalent efficacy and safety established by eculizumab and that patients transition safely and effectively from using eculizumab to ravulizumab. Subsequently, whereas PNH patients may have needed to previously plan their lives rather strictly around the bi-weekly infusion administrations of eculizumab, with ravulizumab such patients can find a more relaxed dosing schedule of only six or seven infusions over an entire year, Label.Specifications
Ravulizumab | |
Recombinant Monoclonal | |
Unconjugated | |
ALXN-1210; ravulizumab-cwvz | |
Human C5 | |
100 μg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
ELISA, SDS-Page | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
Human | |
Protein A | |
RUO | |
Chemical | |
Antibody | |
IgG2 κ, IgG4 κ |