missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rat SREBP2 (aa 843-857) Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP275
This item is not returnable.
View return policy
Description
Rat SREBP2 (residues 843-857) Synthetic Peptide, PEP-275, from Invitrogen.
PEP-275 is a 15 amino acid synthetic peptide whose sequence corresponds to residues 843-857 of rat SREBP 2. The sequence of this peptide is (amino to carboxy terminus): A ( 843)-I-S-W-L-Q-G-D-D-A-A-V-R-S-H (857)This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product, SREBP 2, catalog # PA1-338. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.Specifications
300095 | |
purified | |
Rat SREBP2 (aa 843-857) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Rat | |
Q3T1I5 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
Srebf2 | |
No preservative | |
50 μg | |
A( 843) - I - S - W - L - Q - G - D - D - A - A -V - R - S - H(857) |