missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rat IL-6 Uncoated ELISA Kit with Plates
Specifications
Assay Range | 31.2 to 2,000 pg/mL |
---|---|
Assay Sensitivity | <31.2 pg/mL |
Conjugate | Biotin |
Product Type | ELISA |
Sample Type | Plasma, Serum, Supernatant |
Description
Interleukin 6 (IL-6) is a multifunctional 26 kD protein originally discovered in the medium of RNA-stimulated fibroblastoid cells. IL-6 appears to be directly involved in the responses that occur after infection and cellular injury, and it may prove to be important as IL-1 and TNF-a in regulating the acute phase response. IL-6 is reported to be produced by fibroblasts, activated T cells, activated monocytes or macrophages and endothelial cells. IL-6 acts upon a variety of cells including myeloid progenitor cells, T cells, B cells, and hepatocytes. In addition, IL-6 appears to interact with IL-2 in the proliferation of T lymphocytes. IL-6 potentiates the proliferative effect of IL-3 on multipotential hematopoietic progenitors. IL-6 plays a critical role in B-cell differentiation to plasma cells and is a potent growth factor for plasmacytoma and myeloma. IL-6 is a very useful culture supplement for the generation of a high number of antibody-producing hybridomas. Primarily produced at sites of acute and chronic inflammation, IL-6 is secreted into the serum and induces a transcriptional inflammatory response through interleukin 6 receptor, alpha. The functioning of IL-6 is implicated in a wide variety of inflammation-associated disease states including diabetes mellitus and systemic juvenile rheumatoid arthritis.Specifications
31.2 to 2,000 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
B-cell differentiation factor; B-cell hybridoma growth factor; B-cell stimulatory factor 2; BSF2; BSF-2; CDF; ChIL-6; CTL differentiation factor; cytokine; HGF; H-IL-6; HSF; Hybridoma growth factor; Ifnb2; IFN-beta-2; IL6; Il-6; ILg6; ILN; Interferon beta-2; Interleukin; interleukin 6; interleukin 6 (interferon, beta 2); interleukin 6 precursor; interleukin BSF-2; Interleukin HP-1; Interleukin6; Interleukin-6; interleukin-6 precursor; interleukin-6 protein; M-IL-6; prointerleukin 6; R-IL-6 | |
Il6 | |
HRP | |
Supernatant, 50 μL; Serum, 50 μL; Plasma, 50 μL | |
IL-6 | |
24 hr 30 min |
<31.2 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
P20607 | |
Capture Antibody: Pre-titrated, purified antibody Detection Antibody: Pre-titrated, biotin-conjugated antibody Standard: Recombinant protein for generating standard curve and calibrating samples 10 x Coating Buffer: Buffer for plating the Capture Antibody Assay Buffer A Enzyme: Pre-titrated Streptavidin-HRP Concentrate (100 X) Substrate: 1 x TMB Solution Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards 96 Well Plate: Corning Costar 9018 (included with product Cat. Nos. ending in suffixes -22, -76, -86) |
|
Research Use Only | |
2°C to 8°C | |
1 hr 45 min |