missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Rat Decorin ELISA Kit
Rat Decorin quantitates rat Decorin in serum, plasma, supernatant.
5390.00 NOK - 26530.00 NOK
Specifications
Accession Number | bone proteoglycan II; CSCD; DC; Dcn; DCN protein; DCN1C; Decorin; decorin isoform a preproprotein; Dermatan sulfate proteoglycan-II; dermatan sulfate proteoglycan-II (decorin); dermatan sulphate proteoglycans II; DSPG; DSPG2; mDcn; PG40; PGII; PGS2; PG-S2; proteoglycan core protein; proteoglycan core protein; binds type I collagen fibers and TGF-beta; decoron; similar to bone proteoglycan II; similar to decorin; SLRR1B; small leucine-rich protein 1 B; unnamed protein product |
---|---|
Assay Range | 20 to 5,000 pg/mL |
Assay Sensitivity | 20 pg/mL |
Conjugate | Biotin |
Product Type | ELISA |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17185899
|
Invitrogen™
ERA8RBX5 |
5 x 96 Tests |
26530.00 NOK
Each |
Estimated Shipment: 03-01-2025
Log in to see stock. |
|||||
17175073
|
Invitrogen™
ERA8RB |
96 Tests |
5390.00 NOK
Pack of 96 |
Estimated Shipment: 06-01-2025
Log in to see stock. |
|||||
Description
Rat Decorin quantitates rat Decorin in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant rat Decorin.
Decorin is a member of the small, leucine-rich proteoglycan (SLRP) family along with biglycan. Found in the interterritorial region of cartilage, decorin interacts with collagen and other matrix proteins to regulate the cartilaginous matrix. It has been shown to interact with fibronectin, thrombospondin, C1q, epidermal growth factor receptor (EGFR), and TGF-b. It is believed that decorin also plays a role in the cell cycle of chondrocytes. The decorin neoepitope fragment has a sequence of PKTLQE, and is generated by MMP-3 or ADAMTS-4 cleavage at Glu154-Leu155 of decorin core protein.Specifications
bone proteoglycan II; CSCD; DC; Dcn; DCN protein; DCN1C; Decorin; decorin isoform a preproprotein; Dermatan sulfate proteoglycan-II; dermatan sulfate proteoglycan-II (decorin); dermatan sulphate proteoglycans II; DSPG; DSPG2; mDcn; PG40; PGII; PGS2; PG-S2; proteoglycan core protein; proteoglycan core protein; binds type I collagen fibers and TGF-beta; decoron; similar to bone proteoglycan II; similar to decorin; SLRR1B; small leucine-rich protein 1 B; unnamed protein product | |
20 pg/mL | |
ELISA | |
Colorimetric Microplate Reader | |
DCN | |
<10% | |
HRP | |
Plasma, 50 μL; Supernatant, 50 μL; Serum, 50 μL | |
Decorin | |
4 hr 45 min |
20 to 5,000 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
Q01129 | |
<12% | |
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
Research Use Only | |
2°C to 8°C | |
1 hr 20 min |