Learn More
Sonic Hedgehog/Shh N-Terminus Rat anti-Mouse, Clone: 171018, R&D Systems™
Rat Monoclonal Antibody has been used in 2 publications
Brand:  R&D Systems MAB4641-SP
Encompass
Product Code. 15747034
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Sonic Hedgehog/Shh Monoclonal specifically detects Sonic Hedgehog/Shh in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
Show More
Documents
One moment while we fetch your results.
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Sonic Hedgehog/Shh N-Terminus Rat anti-Mouse, Clone: 171018, R&D Systems™