Learn More
RASIP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 17971-1-AP-20UL
 View more versions of this product
Product Code. 16897984
  • 1696.32 NOK / 20µL
Explore more special offers
Description
Description
Required for the proper formation of vascular structures that develop via both vasculogenesis and angiogenesis. Acts as a critical and vascular-specific regulator of GTPase signaling, cell architecture, and adhesion, which is essential for endothelial cell morphogenesis and blood vessel tubulogenesis. Regulates the activity of Rho GTPases in part by recruiting ARHGAP29 and suppressing RhoA signaling and dampening ROCK and MYH9 activities in endothelial cells. May act as effector for Golgi-bound HRAS and other Ras-like proteins. May promote HRAS-mediated transformation. Negative regulator of amino acid starvation-induced autophagy.
Specifications
Specifications
RASIP1
Polyclonal
Unconjugated
RASIP1
RAIN, Ras interacting protein 1, RASIP1
Rabbit
Antigen Affinity Chromatography
RUO
292912, 54922, 69903
-20°C
Liquid
Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q3U0S6, Q5U651
RASIP1
RASIP1 Fusion Protein Ag11982
20 μL
Primary
Human, Mouse, Rat