missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAP2B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16266-1-AP-20UL
This item is not returnable.
View return policy
Description
This intronless gene belongs to a family of RAS-related genes. The proteins encoded by these genes share approximately 50% amino acid identity with the classical RAS proteins and have numerous structural features in common. The most striking difference between the RAP and RAS proteins resides in their 61st amino acid: glutamine in RAS is replaced by threonine in RAP proteins. Evidence suggests that this protein may be polyisoprenylated and palmitoylated.Specifications
RAP2B | |
Polyclonal | |
Unconjugated | |
RAP2B | |
RAP2B, Ras related protein Rap 2b | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
170923, 5912, 74012 | |
-20°C | |
Liquid |
Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P61225, P61226, P61227 | |
Rap2b | |
RAP2B Fusion Protein Ag9331 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |