Learn More
Novacyt - Microgen Bioproducts Rap-ID L. Mono
Shop All Novacyt Group Products
Click to view available options
Quantity:
96 Tests
Unit Size:
96 tests
Description
- Biochemical identification kit.
- For rapid confirmation of L.monocytogenes isolated on chromogenic agar plate media.
- Complete confirmation system for the identification of L.monocytogenes from chromogenic agar.
Specifications
Specifications
Detectable Analytes | L. monocytogenes |
Type | Biochemical Identification Kit |
For Use With (Application) | Rapid Confirmation of L.Monocytogenes Isolated on Chromogenic Agar Plate Media. |
Quantity | 96 Tests |
Product Content Correction
Product Title
Novacyt - Microgen Bioproducts Rap-ID L. Mono >