Learn More
Fisherbrand™ Gamma Radiation Storage Boxes
Uniquely designed Cleaver Scientific storage boxes are manufactured with hinged lids and accommodate interchangeable inserts to hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes. Ideal solution for short-term storage of radioactive waste or radioisotopes
Brand:  Fisherbrand™ CSR-STOREG
Product Code. 12397197
  • 19190.00 NOK / Each
Only null left
Estimated Shipment: 21-11-2025
Description
  • For storage and transport solid acrylic for 24 x 1.5mL and 15 x 0.5mL tubes 150mm x 120mm x 70mm external Fisherbrand
  • Cleaver Scientific storage boxes are manufactured with hinged lids and accommodate interchangeable inserts to hold microtubes, centrifuge tubes, scintillation vials, universals, cryotubes and falcon tubes
  • Also available is our range of floor-standing and benchtop bins with anti-slip feet and hinged lids, which serve as an ideal solution for short-term storage of radioactive waste or radioisotopes
  • Both the Beta and Gamma storage bins are available in five sizes, and the two largest bin models have wheels for easy transportation
  • Optional heavy duty drawstring bags may also be purchased
  • Gamma Storage/Transport Block (7 x 15 x 12cm)
Specifications
Specifications
7 x 15 x 12cm
Optional heavy duty drawstring bags may also be purchased
Storage boxes for hazard and radiation materials
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Fisherbrand™ Gamma Radiation Storage Boxes > Gamma Storage/Transport Block: (7 x 15 x 12cm)