missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAD17 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13358-1-AP-150UL
This item is not returnable.
View return policy
Description
Rad17 is essential for sustained cell growth, maintece of chromosomal stability, and ATR-dependent checkpoint activation upon DNA damage. It has a weak ATPase activity required for binding to chromatin. Rad17 also participates in the recruitment of the RAD1-RAD9-HUS1 complex and RHNO1 onto chromatin, and in CHEK1 activation. May also serve as a sensor of DNA replication progression, and may be involved in homologous recombination.Specifications
RAD17 | |
Polyclonal | |
Unconjugated | |
RAD17 | |
CCYC, HRAD17, R24L, RAD17, RAD17 homolog (S. pombe), RAD17SP, RAD24, RF C/activator 1 homolog | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19356, 310034, 5884 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O75943, Q6NXW6 | |
Rad17 | |
RAD17 Fusion Protein Ag4171 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |